Skip to Content
Merck
All Photos(1)

Documents

W370703

Sigma-Aldrich

Methyl 2-methylpentanoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
3707
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.549

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.403 (lit.)

bp

45 °C/0.2 mmHg (lit.)

density

0.878 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CCCC(C)C(=O)OC

InChI

1S/C7H14O2/c1-4-5-6(2)7(8)9-3/h6H,4-5H2,1-3H3

InChI key

ZTULNMNIVVMLIU-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service