Skip to Content
Merck
All Photos(1)

Documents

W347205

Sigma-Aldrich

Thiogeraniol

Synonym(s):

3,7-Dimethyl-2,6-octadiene-1-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18S
CAS Number:
Molecular Weight:
170.31
MDL number:
UNSPSC Code:
12164502
Flavis number:
12.064

biological source

synthetic

SMILES string

C\C(C)=C\CC\C(C)=C\CS

InChI

1S/C10H18S/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

FACAUSJJVBMWLV-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service