Key Documents
TEM-CF200F1CU
Sigma-Aldrich
Carbon Film Supported Copper Gilder Finder F1 Grids
size 200 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON GILDER FI FINDER 200# CU 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported glider finder (F1) grids. Copper grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F1 type of design has thick bars dividing regions into 6 thin bar areas, which are identified by a numeric system.
Choose from one of the most recent versions:
Certificates of Analysis (COA)