SYNF100003
Synthware™ 47 mm filtration apparatus with fritted support
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F100003
Related Categories
General description
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Legal Information
Synthware is a trademark of Kemtech America Inc.