Documents
SYNA621922
Synthware vacuum take-off adapter with short stem
joint: ST/NS 19/22, hose size 8 mm
Synonym(s):
Synthware A621922, distillation adapter, vacuum take off adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A621922
hose size
8 mm
Related Categories
General description
With a hose connection and a drip tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.