Skip to Content
Merck
All Photos(1)

Key Documents

RNI00021

Sigma-Aldrich

4-Hydroxyphenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12BNO5
Molecular Weight:
249.03
UNSPSC Code:
12352108
PubChem Substance ID:

form

solid

SMILES string

O=C(CN(C)CC(O1)=O)OB1C2=CC=C(O)C=C2

InChI

1S/C11H12BNO5/c1-13-6-10(15)17-12(13,18-11(16)7-13)8-2-4-9(14)5-3-8/h2-5,14H,6-7H2,1H3

InChI key

WUVDNEDXFYTLIX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service