M2627
5-Methylindole-2-carboxylic acid
Synonym(s):
NSC 88873
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
SMILES string
Cc1ccc2[nH]c(cc2c1)C(O)=O
InChI
1S/C10H9NO2/c1-6-2-3-8-7(4-6)5-9(11-8)10(12)13/h2-5,11H,1H3,(H,12,13)
InChI key
DAITVOCMWPNFTL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
reactant in preparation of Pin1 inhibitors as potential antitumor agents
reactant in preparation of non-imidazole human histamine H4 receptor antagonists
reactant in preparation of non-imidazole human histamine H4 receptor antagonists
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Andrew Potter et al.
Bioorganic & medicinal chemistry letters, 20(22), 6483-6488 (2010-10-12)
Pin1 is an emerging oncology target strongly implicated in Ras and ErbB2-mediated tumourigenesis. Pin1 isomerizes bonds linking phospho-serine/threonine moieties to proline enabling it to play a key role in proline-directed kinase signalling. Here we report a novel series of Pin1
Jill A Jablonowski et al.
Journal of medicinal chemistry, 46(19), 3957-3960 (2003-09-05)
Following the discovery of the human histamine H4 receptor, a high throughput screen of our corporate compound collection identified compound 6 as a potential lead. Investigation of the SAR resulted in the discovery of novel compounds 10e and 10l, which
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service