GF43195147
Polymethylpentene
TPX®, sheet, thickness 22.5 mm, size 150 × 150 mm, natural
Synonym(s):
ME313435
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
sheet
manufacturer/tradename
Goodfellow 431-951-47
size
150 × 150 mm
thickness
22.5 mm
color
natural
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service