Skip to Content
Merck
All Photos(2)

Documents

E18905

Sigma-Aldrich

Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate

99%, solid

Synonym(s):

Ethyl trans-α-cyano-3-indoleacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate, 99%

Assay

99%

form

powder

color

yellow

mp

164-166 °C (dec.) (lit.)

solubility

methanol: 2.5%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCOC(=O)\C(=C\c1c[nH]c2ccccc12)C#N

InChI

1S/C14H12N2O2/c1-2-18-14(17)10(8-15)7-11-9-16-13-6-4-3-5-12(11)13/h3-7,9,16H,2H2,1H3/b10-7+

InChI key

XDRLIASMKGKPDX-JXMROGBWSA-N

General description

Example of a cyanovinylheteroaromatic for organic nonlinear optics.

Application

NLO material

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service