Skip to Content
Merck
All Photos(1)

Documents

DWKWGF538P

Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture

label, Methanol, low-density polyethylene bottle, capacity 500 mL

Synonym(s):

WGF538P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle

feature

label Methanol

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGF538P

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for methannol dispensing within the laboratory

Features and Benefits

Slim design for comfortable handling and ease of use

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service