Key Documents
DWK227494-00G
Wheaton® Bod Bottle
capacity 60 mL, bottle diam. × H 43 mm × 115 mm, pennyhead glass cap (stopper)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
pennyhead glass cap (stopper)
packaging
pkg of 36 ea
manufacturer/tradename
WHEATON® 227494-00G
bottle diam. × H
43 mm × 115 mm
capacity
60 mL
General description
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Features and Benefits
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)