Skip to Content
Merck
All Photos(2)

Key Documents

D166006

Sigma-Aldrich

2,5-Dimethylindole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N
CAS Number:
Molecular Weight:
145.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

mp

112-113 °C (lit.)

SMILES string

Cc1ccc2[nH]c(C)cc2c1

InChI

1S/C10H11N/c1-7-3-4-10-9(5-7)6-8(2)11-10/h3-6,11H,1-2H3

InChI key

ZFLFWZRPMDXJCW-UHFFFAOYSA-N

Application

  • Reactant in stereoselective preparation of substituted indoles via rhodium-catalyzed enantioselective coupling reaction of indoles with diazo compounds
  • Reactant in stereoselective synthesis of indolines via palladium-catalyzed asymmetric hydrogenation of unprotected indoles
  • Reactant in enantioselective synthesis of fluorene derivatives via Friedel-Crafts reactions
  • Reactant in preparation of bis-indolyldihydroxybenzoquinones by addition indoles to dichlorobenzoquinone promoted by Bronsted acid
  • Reactant in reaction with hydroxypyrazolines

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service