Skip to Content
Merck
All Photos(1)

Key Documents

C97601

Sigma-Aldrich

Cyclododecene, mixture of cis and trans

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22
CAS Number:
Molecular Weight:
166.30
Beilstein:
2037841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

232-245 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

C1CCCCCC=CCCCC1

InChI

1S/C12H22/c1-2-4-6-8-10-12-11-9-7-5-3-1/h1-2H,3-12H2/b2-1-

InChI key

HYPABJGVBDSCIT-UPHRSURJSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service