Skip to Content
Merck
All Photos(1)

Key Documents

C97458

Sigma-Aldrich

Cyclododecanone

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H22(=O)
CAS Number:
Molecular Weight:
182.30
Beilstein:
508621
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

bp

85 °C/1 mmHg (lit.)

mp

59-61 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCCC1

InChI

1S/C12H22O/c13-12-10-8-6-4-2-1-3-5-7-9-11-12/h1-11H2

InChI key

SXVPOSFURRDKBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Qiu et al.
Carbohydrate polymers, 182, 75-80 (2017-12-28)
In comparison with natural α- and β-cyclodextrin (CD), γ-CD has attracted much attention due to their large hydrophobic cavities, high water solubility, and bioavailability. However, the production of γ-CD is still rather expensive and time-consuming. To overcome the high cost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service