Skip to Content
Merck
All Photos(1)

Documents

BR705129

Wall mount for HandyStep® S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121506
NACRES:
NB.01

material

PC

packaging

pack of 1 ea

manufacturer/tradename

BRAND 705129

Related Categories

General description

  • For use with HandyStep® S
  • Accessory
  • Storage rack
  • Holds pipetters

Legal Information

HandyStep is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service