Skip to Content
Merck
All Photos(1)

Key Documents

94645

Sigma-Aldrich

L-Valine allyl ester p-toluenesulfonate salt

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)COOCH2CH=CH2 · C7H8O3S
CAS Number:
Molecular Weight:
329.41
Beilstein:
5200860
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99.0%

form

crystals

optical activity

[α]20/D +5.5±1°, c = 1% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

117-120 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)[C@H]([NH3+])C(=O)OCC=C.Cc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C8H15NO2.C7H8O3S/c1-4-5-11-8(10)7(9)6(2)3;1-6-2-4-7(5-3-6)11(8,9)10/h4,6-7H,1,5,9H2,2-3H3;2-5H,1H3,(H,8,9,10)/t7-;/m0./s1

InChI key

HSIRKDASFUCGOZ-FJXQXJEOSA-N

Other Notes

Used in peptide and N-glycopeptide synthesis; the ester is selectively deblocked with (Ph3P)3RhCl[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Waldmann et al.
Liebigs Ann. Chem., 1712-1712 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service