Skip to Content
Merck
All Photos(1)

Key Documents

900947

Sigma-Aldrich

ITIC-M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H86N4O2S4
Molecular Weight:
1456.00
UNSPSC Code:
12352101
Pricing and availability is not currently available.

description

Band gap: 1.6 eV

Assay

≥97%

form

solid

solubility

chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.58 eV 
LUMO -3.98 eV 

SMILES string

O=C1C2=C(C=CC=C2)C(/C1=C/C3=CC(S4)=C(S3)C5=C4C(C=C(C(C6=CC=C(C=C6)CCCCCC)(C7=CC=C(C=C7)CCCCCC)C8=C9SC%10=C8SC(/C=C%11C(C(C=CC=C%12)=C%12C/%11=O)=C(C#N)C#N)=C%10)C9=C%13)=C%13C5(C%14=CC=C(C=C%14)CCCCCC)C%15=CC=C(C=C%15)CCCCCC)=C(C#N)C#N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunsun Li et al.
Advanced materials (Deerfield Beach, Fla.), 28(42), 9423-9429 (2016-09-09)
Fine energy-level modulations of small-molecule acceptors (SMAs) are realized via subtle chemical modifications on strong electron-withdrawing end-groups. The two new SMAs (IT-M and IT-DM) end-capped by methyl-modified dicycanovinylindan-1-one exhibit upshifted lowest unoccupied molecular orbital (LUMO) levels, and hence higher open-circuit

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service