Skip to Content
Merck
All Photos(1)

Documents

857769

Sigma-Aldrich

Ethyl linoleate

98%

Synonym(s):

Linoleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)2(CH2)7COOC2H5
CAS Number:
Molecular Weight:
308.50
Beilstein:
1727827
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.455 (lit.)

bp

224 °C/17 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h8-9,11-12H,3-7,10,13-19H2,1-2H3/b9-8-,12-11-

InChI key

FMMOOAYVCKXGMF-MURFETPASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ethyl linoleate solated from Oxalis triangularis inhibits forskolin-induced melanogenesis and tyrosinase activity in mouse B16 melanoma cells . This anti-melanogenic effect is mediated by inhibiting cAMP production.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service