Skip to Content
Merck
All Photos(1)

Key Documents

779601

Sigma-Aldrich

3-Iodo-2-methylpropene

≥97.0% (GC)

Synonym(s):

1,2-Difluoro-4-iodobenzene, Methallyl iodide, Methylallyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7I
CAS Number:
Molecular Weight:
182.00
Beilstein:
1098356
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

contains

copper as stabilizer

density

1.693 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC(=C)CI

InChI

1S/C4H7I/c1-4(2)3-5/h1,3H2,2H3

InChI key

HVSJHYFLAANPJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.5 °F

Flash Point(C)

35.86 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service