Skip to Content
Merck
All Photos(1)

Documents

766216

Sigma-Aldrich

2-[(4S)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-N-{2-[(4S)-4-(1,1-dimethylethyl)-4,5-dihydro-2-oxazolyl]phenyl}benzenamine

95%

Synonym(s):

non-C2-symmetric bis(oxazoline) ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31N3O2
CAS Number:
Molecular Weight:
405.53
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: ligand

mp

92-96 °C

SMILES string

CC(C)[C@H]1COC(=N1)c2ccccc2Nc3ccccc3C4=N[C@H](CO4)C(C)(C)C

InChI

1S/C25H31N3O2/c1-16(2)21-14-29-23(27-21)17-10-6-8-12-19(17)26-20-13-9-7-11-18(20)24-28-22(15-30-24)25(3,4)5/h6-13,16,21-22,26H,14-15H2,1-5H3/t21-,22-/m1/s1

InChI key

FIJNPJIMVUCDHH-FGZHOGPDSA-N

Application

Ligand for Chromium-Catalyzed Homoallenylation of Aldehydes.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angewandte Chemie (International Edition in English), 48, 1-5 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service