Skip to Content
Merck
All Photos(1)

Key Documents

757314

Sigma-Aldrich

9,9-Dimethyl-N,N′-di(1-naphthyl)-N,N′-diphenyl-9H-fluorene-2,7-diamine

98% (HPLC)

Synonym(s):

DMFL-NPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H36N2
CAS Number:
Molecular Weight:
628.80
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

solid

mp

265-270 °C

Orbital energy

HOMO 5.2 eV 
LUMO 2.3 eV 

SMILES string

CC1(C)c2cc(ccc2-c3ccc(cc13)N(c4ccccc4)c5cccc6ccccc56)N(c7ccccc7)c8cccc9ccccc89

InChI

1S/C47H36N2/c1-47(2)43-31-37(48(35-19-5-3-6-20-35)45-25-13-17-33-15-9-11-23-39(33)45)27-29-41(43)42-30-28-38(32-44(42)47)49(36-21-7-4-8-22-36)46-26-14-18-34-16-10-12-24-40(34)46/h3-32H,1-2H3

InChI key

KJEQVQJWXVHKGT-UHFFFAOYSA-N

Application

This material is a common hole transport material for OLED devices

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Da Y.; et al.
Organic Electronics, 10, 1610-1610 (2009)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service