Skip to Content
Merck
All Photos(2)

Documents

750514

Sigma-Aldrich

Carbon nanotube, single-walled

<1% Metal Catalyst, avg. no. of layers, 1

Synonym(s):

SWCNT, SuperPureTubes, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

30% (Metallic)
70% (Semiconducting)

form

solid

feature

avg. no. of layers 1

manufacturer/tradename

NanoIntegris, Inc.

L

0.3-5 μm

impurities

<1% Metal Catalyst

diameter

1.2-1.7 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

Looking for similar products? Visit Product Comparison Guide

General description

PureTubes are produced by arch discharge or plasma torch. The diameter of the semiconducting single walled nanotubes range in 1.2-1.7 nm, whereas the length varies from 100nm to 4μm. These pure nanotubes have less than 1% of catalyst impurity.

Application

These carbon nanotubes are produced via the Arc Discharge process and are purified via density gradient centrifugation.

Research applications include:
  • Electronics†
  • Sensors†
  • Composites
  • Energy Storage
  • Study of Life Science systems†

Preparation Note

Electric Arc Discharge Method

Legal Information

Product of NanoIntegris, Inc
PureTubes is a trademark of NanoIntegris, Inc.
SuperPureTubes is a trademark of NanoIntegris, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuan Wang et al.
ACS nano, 4(12), 7123-7132 (2010-11-11)
Macroelectronic integrated circuits are widely used in applications such as flat panel display and transparent electronics, as well as flexible and stretchable electronics. However, the challenge is to find the channel material that can simultaneously offer low temperature processing, high
Chad D Vecitis et al.
ACS nano, 4(9), 5471-5479 (2010-09-04)
Single-walled carbon nanotubes (SWNTs) have been previously observed to be strong antimicrobial agents, and SWNT coatings can significantly reduce biofilm formation. However, the SWNT antimicrobial mechanism is not fully understood. Previous studies on SWNT cytotoxicity have concluded that membrane stress
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out

Articles

We presents an article concerning the applications of high-purity single-walled nanotubes in electronic and biomedical fields.

We presents an article concerning the applications of high-purity single-walled nanotubes in electronic and biomedical fields.

We presents an article concerning the applications of high-purity single-walled nanotubes in electronic and biomedical fields.

We presents an article concerning the applications of high-purity single-walled nanotubes in electronic and biomedical fields.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service