740160
Diethyl [(benzylideneamino)methyl] phosphonate
96%
About This Item
Assay
96%
form
liquid
reaction suitability
reaction type: C-C Bond Formation
refractive index
n20/D 1.521
density
1.119 g/mL at 25 °C
SMILES string
CCOP(=O)(C\N=C\c1ccccc1)OCC
InChI
1S/C12H18NO3P/c1-3-15-17(14,16-4-2)11-13-10-12-8-6-5-7-9-12/h5-10H,3-4,11H2,1-2H3/b13-10+
InChI key
REXOGTNRQYAOQG-JLHYYAGUSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service