Skip to Content
Merck
All Photos(1)

Documents

726362

Sigma-Aldrich

4-Bromobenzyl chloride

≥97.0% (GC)

Synonym(s):

1-Bromo-4-(chloromethyl)benzene, p-Bromo-α-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrCl
CAS Number:
Molecular Weight:
205.48
Beilstein:
636506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (GC)

form

crystals

mp

37-38 °C (lit.)

SMILES string

ClCc1ccc(Br)cc1

InChI

1S/C7H6BrCl/c8-7-3-1-6(5-9)2-4-7/h1-4H,5H2

InChI key

BSIIGUGKOPPTPZ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelie Juin et al.
Developmental cell, 51(4), 431-445 (2019-11-02)
Pancreatic ductal adenocarcinoma is one of the most invasive and metastatic cancers and has a dismal 5-year survival rate. We show that N-WASP drives pancreatic cancer metastasis, with roles in both chemotaxis and matrix remodeling. lysophosphatidic acid, a signaling lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service