Skip to Content
Merck
All Photos(1)

Key Documents

720038

Sigma-Aldrich

N-[2-(Diphenylphosphino)benzylidene]cyclohexylamine

97%

Synonym(s):

[2-[(Cyclohexylimino)methyl]phenyl]diphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26NP
CAS Number:
Molecular Weight:
371.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

116-120 °C

functional group

phosphine

SMILES string

C1(P(C2=CC=CC=C2)C3=CC=CC=C3/C=N/C4CCCCC4)=CC=CC=C1

InChI

1S/C25H26NP/c1-4-13-22(14-5-1)26-20-21-12-10-11-19-25(21)27(23-15-6-2-7-16-23)24-17-8-3-9-18-24/h2-3,6-12,15-20,22H,1,4-5,13-14H2/b26-20+

InChI key

CWYDMJZWNROFGL-LHLOQNFPSA-N

Application

Catalyst for:
  • Cross-coupling reactions
  • Stannylative cycloaddition of conjugated enynes
  • Stereo- and regioselective alkynylstannylation of alkynes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service