Skip to Content
Merck
All Photos(1)

Documents

716723

Sigma-Aldrich

Zinc

shot, 99.9999%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.9999%

form

shot

reaction suitability

reagent type: catalyst
core: zinc

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

Zinc is a high purity precursor for the preparation Zn-based films (PVD) or materials like ZnTe.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
A S Hussein et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 14(2), 113-118 (2013-06-14)
To assess the salivary levels of Copper (Cu), Zinc (Zn), Manganese (Mn) and Iron (Fe) obtained from children of different ethnic backgrounds in Shah Alam, Malaysia and investigate the possible relationships with caries. One hundred and twenty primary school children
A viewpoint about the treatment of Wilson's disease.
Abdul Qayyum Rana et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 612-614 (2013-06-22)
Nalini Pandey et al.
Indian journal of experimental biology, 51(7), 548-555 (2013-08-01)
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service