Skip to Content
Merck
All Photos(1)

Key Documents

710016

Sigma-Aldrich

2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane solution

0.5 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39N4P
CAS Number:
Molecular Weight:
342.50
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

concentration

0.5 M in diethyl ether

density

0.766 g/mL at 25 °C

SMILES string

CC(C)CN1CCN2CCN(CC(C)C)P1N(CC2)CC(C)C

InChI

1S/C18H39N4P/c1-16(2)13-20-10-7-19-8-11-21(14-17(3)4)23(20)22(12-9-19)15-18(5)6/h16-18H,7-15H2,1-6H3

InChI key

WFHPXSHLCFHEIA-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service