Skip to Content
Merck
All Photos(1)

Key Documents

707694

Sigma-Aldrich

3-(Di-tert-butylphosphonium)propane sulfonate

97%

Synonym(s):

DTBPPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H25O3PS
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

132 °C

functional group

phosphine

SMILES string

CC(C)(C)P(CCCS(O)(=O)=O)C(C)(C)C

InChI

1S/C11H25O3PS/c1-10(2,3)15(11(4,5)6)8-7-9-16(12,13)14/h7-9H2,1-6H3,(H,12,13,14)

InChI key

JPNPRWMRUCIEMN-UHFFFAOYSA-N

Application

Air stable, water soluble phosphine ligand used in palladium catalyzed cross-coupling reactions

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brown, W. S. et al.
Synthesis, 1965-1965 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service