Skip to Content
Merck
All Photos(1)

Key Documents

695793

Sigma-Aldrich

Ethyl 4-(4-benzyloxyphenyl)-2,4-dioxobutanoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O5
CAS Number:
Molecular Weight:
326.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

123-128 °C

SMILES string

CCOC(=O)C(=O)CC(=O)c1ccc(OCc2ccccc2)cc1

InChI

1S/C19H18O5/c1-2-23-19(22)18(21)12-17(20)15-8-10-16(11-9-15)24-13-14-6-4-3-5-7-14/h3-11H,2,12-13H2,1H3

InChI key

BTUJZIGPAGODTR-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service