Skip to Content
Merck
All Photos(1)

Key Documents

679577

Sigma-Aldrich

Methyl 3,5-dichloropyridine-4-carboxylate

97%

Synonym(s):

3,5-Dichloropyridine-4-carboxylic acid methyl ester, Methyl 3,5-dichloroisonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NO2
CAS Number:
Molecular Weight:
206.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.5362

bp

110 °C/1 mmHg

density

1.244 g/mL at 25 °C

SMILES string

COC(=O)c1c(Cl)cncc1Cl

InChI

1S/C7H5Cl2NO2/c1-12-7(11)6-4(8)2-10-3-5(6)9/h2-3H,1H3

InChI key

DVLQQMUAXUCZGE-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service