Skip to Content
Merck
All Photos(1)

Key Documents

667773

Sigma-Aldrich

α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CF3)(OCH3)COCl
CAS Number:
Molecular Weight:
252.62
Beilstein:
2215441
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.471 (lit.)

bp

213-214 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(C(Cl)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H8ClF3O2/c1-16-9(8(11)15,10(12,13)14)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

PAORVUMOXXAMPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

190.0 °F

Flash Point(C)

87.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service