Skip to Content
Merck
All Photos(1)

Key Documents

657506

Sigma-Aldrich

3-[(4′-(2-Methoxyethyl)phenoxy)methyl]phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19BO4
CAS Number:
Molecular Weight:
286.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

mp

98-104 °C

SMILES string

COCCc1ccc(OCc2cccc(c2)B(O)O)cc1

InChI

1S/C16H19BO4/c1-20-10-9-13-5-7-16(8-6-13)21-12-14-3-2-4-15(11-14)17(18)19/h2-8,11,18-19H,9-10,12H2,1H3

InChI key

RABCYEGFKLWONK-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service