Skip to Content
Merck
All Photos(2)

Documents

650625

Sigma-Aldrich

4-Amino-2-fluorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(F)CO2H
CAS Number:
Molecular Weight:
155.13
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

210 °C (dec.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(C(O)=O)c(F)c1

InChI

1S/C7H6FNO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,9H2,(H,10,11)

InChI key

QHERSCUZBKDVOC-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Bin Luo et al.
ACS nano, 13(7), 8417-8424 (2019-07-04)
Organic color center-tailored semiconducting single-walled carbon nanotubes are a rising family of synthetic quantum emitters that display bright defect photoluminescence molecularly tunable for imaging, sensing, and quantum information processing. A major advance in this area would be the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service