Skip to Content
Merck
All Photos(1)

Key Documents

648922

Sigma-Aldrich

2-(4-Fluorophenyl)-3-butyn-2-ol

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4C(CH3)(OH)C≡CH
CAS Number:
Molecular Weight:
164.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

90%

refractive index

n20/D 1.514

density

1.111 g/mL at 25 °C

functional group

fluoro
hydroxyl

storage temp.

2-8°C

SMILES string

CC(O)(C#C)c1ccc(F)cc1

InChI

1S/C10H9FO/c1-3-10(2,12)8-4-6-9(11)7-5-8/h1,4-7,12H,2H3

InChI key

JEHIMMYBAYPVHN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service