Skip to Content
Merck
All Photos(1)

Key Documents

639540

Sigma-Aldrich

3-Chloro-4-(3′-chlorobenzyloxy)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BCl2O3
CAS Number:
Molecular Weight:
296.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

180-185 °C (lit.)

SMILES string

OB(O)c1ccc(OCc2cccc(Cl)c2)c(Cl)c1

InChI

1S/C13H11BCl2O3/c15-11-3-1-2-9(6-11)8-19-13-5-4-10(14(17)18)7-12(13)16/h1-7,17-18H,8H2

InChI key

HGXKMYLFVPMNFR-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service