Skip to Content
Merck
All Photos(1)

Key Documents

63403

Sigma-Aldrich

Ethyl malonate monoamide

≥95.0% (HPLC)

Synonym(s):

Ethyl carbamoylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COCH2COOCH2CH3
CAS Number:
Molecular Weight:
131.13
Beilstein:
1758870
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

solid

mp

47-50 °C

SMILES string

CCOC(=O)CC(N)=O

InChI

1S/C5H9NO3/c1-2-9-5(8)3-4(6)7/h2-3H2,1H3,(H2,6,7)

InChI key

UVSPVEYCSVXYBB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service