Skip to Content
Merck
All Photos(2)

Key Documents

595837

Sigma-Aldrich

(S)-(−)-3,3′-Dibromo-1,1′-bi-2-naphthol

96%

Synonym(s):

(S)-3,3′-Dibromo-1,1′-Binaphthalene-2,2′-diol, (S)-3,3′-Dibromo-1,1′-bi-2-naphthol, (S)-3,3′-Dibromo-BINOL, (S)-Dibromo-1,1′-Bi-2,2′-naphthol, (S)-Dibromo-1,1′-Binaphthalene-2,2′-diol, (S)-Dibromo-1,1-Binaphthol, (S)-Dibromo-BINOL, (S)-Dibromo-bi-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12Br2O2
CAS Number:
Molecular Weight:
444.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

solid

mp

256-260 °C

SMILES string

Oc1c(Br)cc2ccccc2c1-c3c(O)c(Br)cc4ccccc34

InChI

1S/C20H12Br2O2/c21-15-9-11-5-1-3-7-13(11)17(19(15)23)18-14-8-4-2-6-12(14)10-16(22)20(18)24/h1-10,23-24H

InChI key

BRTBEAXHUYEXSY-UHFFFAOYSA-N

General description

(S)-(-)-3,3′-Dibromo-1,1′-bi-2-naphthol may be used as a catalyst in the preparation of (R)-1-(2-propenyl)-2-(3-phenoxymethoxypropyl)-2-cyclohexenol.[1]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Conjugate Allylation of Cyclic Enones.
Taber DF, et al.
The Journal of Organic Chemistry, 76(18), 7614-7617 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service