591459
Methyl (3S)-(+)-3-(1-methylindol-3-yl)butanoate
optical purity ee: 97% (HPLC), 98%
About This Item
Assay
98%
optical activity
[α]20/D +7°, c = 1% in chloroform
optical purity
ee: 97% (HPLC)
refractive index
n20/D 1.5590 (lit.)
bp
344-345 °C (lit.)
density
1.093 g/mL at 25 °C (lit.)
SMILES string
COC(=O)C[C@H](C)c1cn(C)c2ccccc12
InChI
1S/C14H17NO2/c1-10(8-14(16)17-3)12-9-15(2)13-7-5-4-6-11(12)13/h4-7,9-10H,8H2,1-3H3/t10-/m0/s1
InChI key
QIOUBQLAFUPETC-JTQLQIEISA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service