Skip to Content
Merck
All Photos(1)

Key Documents

571458

Sigma-Aldrich

1-(2-Phenylethyl)piperazine

97%

Synonym(s):

1-Phenethylpiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2
CAS Number:
Molecular Weight:
190.28
Beilstein:
145893
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5430 (lit.)

bp

276 °C (lit.)

density

>1.006 g/mL at 20 °C (lit.)

SMILES string

C1CN(CCN1)CCc2ccccc2

InChI

1S/C12H18N2/c1-2-4-12(5-3-1)6-9-14-10-7-13-8-11-14/h1-5,13H,6-11H2

InChI key

LKUAPSRIYZLAAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

Air-sensitive - absorbs CO2

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service