Skip to Content
Merck
All Photos(1)

Key Documents

571229

Sigma-Aldrich

(4-(1-Piperidinylmethyl)phenyl)magnesium bromide solution

0.25 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4CH2NC5H10MgBr
CAS Number:
Molecular Weight:
278.47
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.25 M in THF

bp

65 °C

density

0.924 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Mg]c1ccc(CN2CCCCC2)cc1

InChI

1S/C12H16N.BrH.Mg/c1-3-7-12(8-4-1)11-13-9-5-2-6-10-13;;/h3-4,7-8H,2,5-6,9-11H2;1H;/q;;+1/p-1

InChI key

IHERGHRULIMGJW-UHFFFAOYSA-M

Application

(4-(1-Piperidinylmethyl)phenyl)magnesium bromide is a Grignard reagent that can be used:
  • As a substrate in the synthesis of functionalized phenols via aerobic oxidation.
  • To prepare set of rosamine derivatives, which are used as glutathione probes in living cells.

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combinatorial rosamine library and application to in vivo glutathione probe
Ahn Y, et al.
Journal of the American Chemical Society, 129(15), 4510-4511 (2007)
Continuous-Flow Synthesis of Functionalized Phenols by Aerobic Oxidation of Grignard Reagents
He Zhi and Jamison TF
Angewandte Chemie (International Edition in English), 53(13), 3353-3357 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service