All Photos(1)
About This Item
Linear Formula:
F2C6H3SO2NH2
CAS Number:
Molecular Weight:
193.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
157-161 °C (lit.)
SMILES string
NS(=O)(=O)c1cc(F)cc(F)c1
InChI
1S/C6H5F2NO2S/c7-4-1-5(8)3-6(2-4)12(9,10)11/h1-3H,(H2,9,10,11)
InChI key
MKQPOVUFDWKPNO-UHFFFAOYSA-N
General description
3,5-Difluorobenzenesulfonamide is a fluorinated benzenesulfonamide. It is a CA (carbonic anhydrase) inhibitor that binds to human carbonic anhydrase I by forming a 1:1 complex.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fluorine NMR studies of the human carbonic anhydrase-3, 5-difluorobenzenesulfonamide complex
Veenstra DL and Gerig JT
Magnetic Resonance in Chemistry, 36(S1), S169-S178 (1998)
Virginija Dudutienė et al.
Bioorganic & medicinal chemistry, 21(7), 2093-2106 (2013-02-12)
A series of 4-substituted-2,3,5,6-tetrafluorobenezenesulfonamides were synthesized and their binding potencies as inhibitors of recombinant human carbonic anhydrase isozymes I, II, VII, XII, and XIII were determined by the thermal shift assay, isothermal titration calorimetry, and stop-flow CO2 hydration assay. All
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service