Recommended Products
Assay
97%
form
solid
mp
34-38 °C (lit.)
SMILES string
Fc1cc(Cl)ccc1S(Cl)(=O)=O
InChI
1S/C6H3Cl2FO2S/c7-4-1-2-6(5(9)3-4)12(8,10)11/h1-3H
InChI key
ZFZRENUEJCOCRE-UHFFFAOYSA-N
Application
4-Chloro-2-fluorobenzenesulfonyl chloride (4-chloro-2-fluorobenzene-1-sulfonyl chloride) may be used to synthesize [1-(4-chloro-2-fluorophenylsulfonyl) piperidin-4-yl] diphenylmethanol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[1-(4-Chloro-2-fluorophenylsulfonyl) piperidin-4-yl] diphenylmethanol.
Acta Crystallographica Section E, Structure Reports Online, 63(9), o3669-o3669 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service