531154
Poly(D,L-lactide-co-glycolide)
average Mw 5,000-15,000, lactide:glycolide (50:50)
Synonym(s):
PLGA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Recommended Products
feed ratio
lactide:glycolide (50:50)
mol wt
average Mw 5,000-15,000
inherent viscosity
0.15-0.25 dL/g(lit.)
transition temp
Tg (DSC) 24 °C (onset)
SMILES string
OCC(O)=O.CC(O)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.
Physical form
Biocompatible, biodegradable polymer.
comparable product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Claus G Madsen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 1-7 (2015-02-11)
This study describes how protein release from polymer matrices correlate with simple measurements on the intrinsic viscosity of the polymer solutions used for casting the matrices and calculations of the solubility parameters of polymers and solvents used. Matrices of poly(dl-lactide-co-glycolide)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service