Skip to Content
Merck
All Photos(2)

Key Documents

526436

Sigma-Aldrich

2,4,6-Trichlorobenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2CH2OH
CAS Number:
Molecular Weight:
211.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

98-101 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C7H5Cl3O/c8-4-1-6(9)5(3-11)7(10)2-4/h1-2,11H,3H2

InChI key

WKJWKKDGJLKFER-UHFFFAOYSA-N

Related Categories

Application

2,4,6-Trichlorobenzyl alcohol may be used in the enantioselective synthesis of sulfinite ester.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hillary M Peltier et al.
Organic letters, 7(9), 1733-1736 (2005-04-23)
[reaction: see text] Practical reaction conditions for the catalytic enantioselective synthesis of sulfinate esters are reported. Commercially available cinchona alkaloids were found to be superior catalysts for the sulfinyl transfer reaction of tert-butanesulfinyl chloride and a variety of benzyl alcohols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service