Skip to Content
Merck
All Photos(1)

Documents

524271

Sigma-Aldrich

Allyl(cyclopentadienyl)nickel(II)

97%

Synonym(s):

(η3 -Allyl)(η5 -cyclopentadienyl)nickel, Allylcyclopentadienylnickel, Cyclopentadienylallylnickel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10Ni
CAS Number:
Molecular Weight:
164.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

reaction suitability

core: nickel
reagent type: catalyst

density

1.31 g/mL at 25 °C (lit.)

SMILES string

[Ni].[CH2][CH][CH2].[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.C3H5.Ni/c1-2-4-5-3-1;1-3-2;/h1-5H;3H,1-2H2;

InChI key

RKDYEOKQRRKFGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Pyr. Liq. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service