Skip to Content
Merck
All Photos(1)

Documents

498890

Sigma-Aldrich

3-Methylbutylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2ZnBr
CAS Number:
Molecular Weight:
216.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.97 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)CC[Zn]Br

InChI

1S/C5H11.BrH.Zn/c1-4-5(2)3;;/h5H,1,4H2,2-3H3;1H;/q;;+1/p-1

InChI key

NCVCJECJAXGYFE-UHFFFAOYSA-M

Application

3-Methylbutylzinc bromide can be used:
  • To prepare methyl 4-(3-methylbutanoyl)benzoate, a key starting material for the synthesis of indazole-based glucagon receptor antagonists.
  • To synthesize deuterium-labeled azanickelacyclobutane by reacting with deuterium-labeled aziridine and Me4phen/ NiCl2.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and structure activity relationships of indazole and indole derivatives as potent glucagon receptor antagonists
Song F, et al.
Bioorganic & Medicinal Chemistry Letters, 29(15), 1974-1980 (2019)
Highly regioselective nickel-catalyzed cross-coupling of N-tosylaziridines and alkylzinc reagents
Jensen KL, et al.
Journal of the American Chemical Society, 136(31), 11145-11152 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service