Skip to Content
Merck
All Photos(1)

Key Documents

481904

Sigma-Aldrich

5-Fluoro-2-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
185.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

mp

131-134 °C (lit.)

SMILES string

OC(=O)c1cc(F)ccc1[N+]([O-])=O

InChI

1S/C7H4FNO4/c8-4-1-2-6(9(12)13)5(3-4)7(10)11/h1-3H,(H,10,11)

InChI key

GHYZIXDKAPMFCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Fluoro-2-nitrobenzoic acid, an ortho-nitro benzoic acid derivative, can be prepared by the oxidation of 5-fluoro-2-nitrotoluene. It is an intermediate formed during the synthesis of N1-(2,4-dichlorophenyl)-2-amino-5-fluorobenzamide.[1] It participates in the synthesis of novel quinazolinones that are potential inhibitors of p38α mitogen-activated protein kinase (MAPK).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution and solid-phase approaches to quinconazole and fluquinconazole-inhibitors of fungal ergosterol biosynthesis.
Bilokin YV and Kovalenko SM.
Heterocyclic Communications, 6(5), 409-414 (2000)
Dearg S Brown et al.
Bioorganic & medicinal chemistry letters, 22(12), 3879-3883 (2012-05-23)
A novel, potent and selective quinazolinone series of inhibitors of p38α MAP kinase has been identified. Modifications designed to address the issues of poor aqueous solubility and high plasma protein binding as well as embedded aniline functionalities resulted in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service