Skip to Content
Merck
All Photos(1)

Documents

480436

Sigma-Aldrich

Benzyl bromide-d7

98 atom % D

Synonym(s):

α-Bromotoluene-d7

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD2Br
CAS Number:
Molecular Weight:
178.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.574 (lit.)

bp

198-199 °C (lit.)

mp

−3-−1 °C (lit.)

density

1.497 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])Br

InChI

1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D,6D2

InChI key

AGEZXYOZHKGVCM-XZJKGWKKSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service