Skip to Content
Merck
All Photos(1)

Key Documents

478547

Sigma-Aldrich

2-Methoxy-5-methylphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)NCO
CAS Number:
Molecular Weight:
163.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.537 (lit.)

bp

240 °C (lit.)

mp

25-26 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(C)cc1N=C=O

InChI

1S/C9H9NO2/c1-7-3-4-9(12-2)8(5-7)10-6-11/h3-5H,1-2H3

InChI key

IDOHLSFNBKNRRJ-UHFFFAOYSA-N

General description

2-Methoxy-5-methylphenyl isocyanate is an aryl isocyanate.

Application

2-Methoxy-5-methylphenyl isocyanate may be used in the synthesis of 9-benzyl-9-azabicyclo[3.3.1]nonan-3-yl-2-methoxy-5-methylphenylcarbamate, a precursor to develop σ2 receptor ligands.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhua Chu et al.
Bioorganic & medicinal chemistry, 17(3), 1222-1231 (2009-01-03)
A series of N-substituted 9-azabicyclo[3.3.1]nonan-3alpha-yl phenylcarbamate analogs were synthesized. Among them, WC-26 and WC-59 were identified as the most potent sigma(2) receptor ligands (K(i)=2.58 and 0.82 nM, respectively) with high selectivity against sigma(1) (K(i) of sigma(1)/sigma(2) ratio=557 and 2087, respectively).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service