Skip to Content
Merck
All Photos(2)

Key Documents

476358

Sigma-Aldrich

4,4′-(1,1′-Biphenyl-4,4′-diyldioxy)dianiline

97%

Synonym(s):

4,4′-Bis(4-aminophenoxy)biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-C6H4OC6H4NH2)2
CAS Number:
Molecular Weight:
368.43
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

mp

197-200 °C (lit.)

SMILES string

Nc1ccc(Oc2ccc(cc2)-c3ccc(Oc4ccc(N)cc4)cc3)cc1

InChI

1S/C24H20N2O2/c25-19-5-13-23(14-6-19)27-21-9-1-17(2-10-21)18-3-11-22(12-4-18)28-24-15-7-20(26)8-16-24/h1-16H,25-26H2

InChI key

HYDATEKARGDBKU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service